Orders LinkContactUsLinkComponent
Salt bridges
Salt bridges
Replacement salt bridge, AL110, double liquid junction, ceramic junction, 250 mm length, for Ø 7,5 or 8 mm reference electrodes
Katalog # HACHB40A550
Leverandør:   Hach
Salt bridges
Katalog # HACHB40A550
Leverandør:  Hach
Salt bridges
Salt bridges
Replacement salt bridge, AL110, double liquid junction, ceramic junction, 250 mm length, for Ø 7,5 or 8 mm reference electrodes
Price
625,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.
Specifikationer
  • Beskrivelse:
    Replacement salt bridge, AL110, double liquid junction, ceramic junction, 250 mm length, for Ø 7,5 or 8 mm reference electrodes
  • Seq:
    02