Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
Cover glasses, square, without cords and blisters, 20×20 mm, #1
Katalog # KNITVD12020Y100A
Leverandør:   KNITTEL
Cover glasses
Katalog # KNITVD12020Y100A
Leverandør:  KNITTEL
Cover glasses
Cover glasses, square, without cords and blisters, 20×20 mm, #1
Price
575,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
Certifications: ISO 8255/1
Emballering: 100 pieces per box - 1000 pieces per carton.
Documents
Specifikationer
  • Antal:
    #1
  • Beskrivelse:
    Cover glasses, square, without cords and blisters
  • L×B:
    20×20 mm