Screwthread joints for VACUU.LAN®
Elbow (90°) adapter, 43 mm, for 1/8" NPT thread to PTFE tubing 10/8 mm
VACU20639795
Adapters for VACUU.LAN® local area vacuum network components.
- Beskrivelse:Elbow (90°) adapter, 43 mm, for 1/8" NPT thread to PTFE tubing 10/8 mm
- Seq:04