Microplates, 96-well
Hvid, LUMITRAC™ 600 høj binding, steril, 40 - 340 µl, 96, +, 392 µl
Leverandør: Greiner Bio-One
Gå til:
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Documents
Specifikationer
- Ant. brønde:96
- Anbefalet arbejdsvolumen:40 - 340 µl
- Steril:+
- Beskrivelse:Hvid, LUMITRAC™ 600 høj binding, steril
- Seq:11
- Total volumen:392 µl
- Cat. No.:738-0016
- Miljøcertificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lav fremstillingspåvirkning