Orders LinkContactUsLinkComponent
Bulk material, Hydromatrix™
Bulk material, Hydromatrix™
Hydromatrix™ bulk material, 1 kg
Katalog # VARI198003
Leverandør:   VARIAN
Bulk material, Hydromatrix™
Katalog # VARI198003
Leverandør:  VARIAN
CAS nummer:  
Bulk material, Hydromatrix™
Hydromatrix™ bulk material, 1 kg
CAS nummer:  
Price
3.470,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
SLE products use a broad-performance, inert diatomaceous earth sorbent for rapid, general sample preparation.
Specifikationer
Specifikationer
  • Pakn:
    1 kg
  • Beskrivelse:
    Hydromatrix™ bulk material
Specifikationer