Bond Elut adapters
Adapter cap for 12 and 20 ml Bond Elut cartridges
Katalog # VARI12131003
Leverandør: VARIAN
Gå til:
Product Details & Documents
Bond Elut adapters fit on top of any Bond Elut cartridge and contain a female Luer fitting that accommodates the tip of another cartridge.
- Connect SPE cartridges in series for large samples
- Expand cartridge volume for even more applications
- Transfer large-volume samples to any SPE cartridge
Specifikationer
- Beskrivelse:Adapter cap for 12 and 20 ml Bond Elut cartridges