Orders LinkContactUsLinkComponent
Pipette stand, 337
Pipette stand, 337
7 enkeltkanalpipetter, Polyamid, Rosé
Catalog # SOCO320.337R
Supplier:   Socorex Isba
Pipette stand, 337
Catalog # SOCO320.337R
Supplier:  Socorex Isba
Pipette stand, 337
Pipette stand, 337
7 enkeltkanalpipetter, Polyamid, Rosé
Price
757,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
This space-saving pipette stand made from PA holds up to seven single channel pipettes of most brands.
  • Made of sturdy polyamid material, easy to clean
  • Very stable with anti-slip pads
  • Available in five pastel colours
Specifications
  • Til:
    7 enkeltkanalpipetter
  • Farve:
    Rosé
  • Materiale:
    Polyamid