Pipette stand, 337
7 enkeltkanalpipetter, Polyamid, Rosé
Catalog # SOCO320.337R
Supplier: Socorex Isba
Jump to:
Product Details & Documents
This space-saving pipette stand made from PA holds up to seven single channel pipettes of most brands.
- Made of sturdy polyamid material, easy to clean
- Very stable with anti-slip pads
- Available in five pastel colours
Specifications
- Til:7 enkeltkanalpipetter
- Farve:Rosé
- Materiale:Polyamid