Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Magnetic stirring bars, plain, PTFE

Magnetic stirring bars, plain, PTFE

25 mm, 8 mm
Katalog # COWI001.225.8
Leverandør:   COWIE
Magnetic stirring bars, plain, PTFE
Katalog # COWI001.225.8
Leverandør:  COWIE
Magnetic stirring bars, plain, PTFE
Magnetic stirring bars, plain, PTFE

25 mm, 8 mm

Price
280,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.

Specifikationer
  • Ø:
    8 mm
  • Længde:
    25 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.