Orders LinkContactUsLinkComponent
Lead Standard Solutions, NIST
Lead Standard Solutions, NIST
Bly, Pb, 10 mg/L, 25 mL
Katalog # HACH2374820
Leverandør:   Hach
Lead Standard Solutions, NIST
Katalog # HACH2374820
Leverandør:  Hach
CAS nummer:  
Lead Standard Solutions, NIST
Lead Standard Solutions, NIST
Bly, Pb, 10 mg/L, 25 mL
CAS nummer:  
Price
188,00 kr
Log ind for tilgængelighed
1 * 25 mL
Product Details & Documents
Standard solutions for accuracy checks in lead determinations.
Specifikationer
Specifikationer
  • Koncentration:
    10 mg/L
  • Pakn:
    25 mL
  • Element:
    Bly
  • Symbol:
    Pb
Specifikationer