Orders LinkContactUsLinkComponent
Chlorine standard solution
Chlorine standard solution
Chlorine standard solution, 2 ml PourRite ampules (NIST), 25 - 30 mg/l as Cl₂, 20
Katalog # HACH2630020
Leverandør:   Hach
Chlorine standard solution
Katalog # HACH2630020
Leverandør:  Hach
CAS nummer:  
Chlorine standard solution
Chlorine standard solution
Chlorine standard solution, 2 ml PourRite ampules (NIST), 25 - 30 mg/l as Cl₂, 20
CAS nummer:  
Price
688,00 kr
Log ind for tilgængelighed
1 * 20 Items
Product Details & Documents
For accuracy checks in chlorine determinations. Lot specific concentration displayed on label.
  • Simple accuracy checks
  • Minimise waste
  • Helps ensure reliable result
Specifikationer
Specifikationer
  • Koncentration:
    25 - 30 mg/l as Cl₂
  • Pakn:
    20
  • Beskrivelse:
    Chlorine standard solution, 2 ml PourRite ampules (NIST)