Orders LinkContactUsLinkComponent
Spherical ground joints, Rotulex®
joint size 19
Catalog # BIBBRM19
Supplier:   SciLabware
Spherical ground joints, Rotulex®
Catalog # BIBBRM19
Supplier:  SciLabware
Spherical ground joints, Rotulex®
Spherical ground joints, Rotulex®
joint size 19
Price
978,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
Made of borosilicate glass.
  • All glass and PTFE pathway is highly resistant to chemical attack
  • Can be used at temperatures up to 200 °C
  • System comprises male ball joint with PTFE covered 'O' ring which fits into a female cup joint
  • Joint can be retained together with spring loaded metal clip
Rotulex® offers a grease free spherical joint.
 : Component parts should be ordered separately .
Specifications
  • Model:
    joint size 19
  • Seq:
    02