Storage and Presentation Boxes, with Hinged Lid, V5
V5-32, 117×90×34 mm
Catalog # MULC41332
Supplier: MULTIROIR
Jump to:
Product Details & Documents
Manufactured in crystal polystyrene, these boxes feature an excellent transparent clarity and a high quality finish.
All models are available in standard black base and crystal lid, but are also available with a crystal base and lid.
- Ideal for the presentation of products
- Transparent lid and black background for great visibility
- Available in different sizes
Specifications
- Model:V5-32
- Seq:06
- B×D×H:117×90×34 mm