Orders LinkContactUsLinkComponent
Storage and Presentation Boxes, with Hinged Lid, V5
V5-32, 117×90×34 mm
Catalog # MULC41332
Supplier:   MULTIROIR
Storage and Presentation Boxes, with Hinged Lid, V5
Catalog # MULC41332
Supplier:  MULTIROIR
Storage and Presentation Boxes, with Hinged Lid, V5
Storage and Presentation Boxes, with Hinged Lid, V5
V5-32, 117×90×34 mm
Price
37,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Manufactured in crystal polystyrene, these boxes feature an excellent transparent clarity and a high quality finish.
All models are available in standard black base and crystal lid, but are also available with a crystal base and lid.
  • Ideal for the presentation of products
  • Transparent lid and black background for great visibility
  • Available in different sizes
Specifications
  • Model:
    V5-32
  • Seq:
    06
  • B×D×H:
    117×90×34 mm