Storage and Presentation Boxes, with Hinged Lid, V5
V5-26, 74×74×38 mm
Katalog # MULC41326
Leverandør: MULTIROIR
Gå til:
Product Details & Documents
Manufactured in crystal polystyrene, these boxes feature an excellent transparent clarity and a high quality finish.
All models are available in standard black base and crystal lid, but are also available with a crystal base and lid.
- Ideal for the presentation of products
- Transparent lid and black background for great visibility
- Available in different sizes
Specifikationer
- Model:V5-26
- Seq:05
- B×D×H:74×74×38 mm