Orders LinkContactUsLinkComponent
Storage and Presentation Boxes, with Hinged Lid, V5
Storage and Presentation Boxes, with Hinged Lid, V5
V5-26, 74×74×38 mm
Katalog # MULC41326
Leverandør:   MULTIROIR
Storage and Presentation Boxes, with Hinged Lid, V5
Katalog # MULC41326
Leverandør:  MULTIROIR
Storage and Presentation Boxes, with Hinged Lid, V5
Storage and Presentation Boxes, with Hinged Lid, V5
V5-26, 74×74×38 mm
Price
29,60 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Manufactured in crystal polystyrene, these boxes feature an excellent transparent clarity and a high quality finish.
All models are available in standard black base and crystal lid, but are also available with a crystal base and lid.
  • Ideal for the presentation of products
  • Transparent lid and black background for great visibility
  • Available in different sizes
Specifikationer
  • Model:
    V5-26
  • Seq:
    05
  • B×D×H:
    74×74×38 mm