Drawers, Transparent
7 l, transparent, 210×365×125 mm
216-4349
System of modular drawer cabinets made from PS. For well organised storage of many items, on shelves or at a workstation.
- Total visibility in the drawer
- Each drawer can be assembled horizontally and vertically
- Each drawer can divided with separators
- Kapacitet:7 l
- Farve:transparent
- Seq:03
- B×D×H:210×365×125 mm
Accessories
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse