Laboratory trays, coloured
blå, 175×100×34 mm, 205×130×37 mm, 175×100×34 mm
Katalog # 216-1117
Leverandør: MULTIROIR
Gå til:
Product Details & Documents
PS.
- Five different colours
- Two sizes
- Good chemical and temperature resistance
Multipurpose trays for a wide range of applications.
Specifikationer
- B×D×H udv.:205×130×37 mm
- Farve:blå
- Seq:04
- B×D×H:175×100×34 mm
- B×D×H indv.:175×100×34 mm