Orders LinkContactUsLinkComponent
Laboratory trays, coloured
Laboratory trays, coloured
rød, 260×196×42 mm, 210×150×40 mm
Katalog # 216-1130
Leverandør:   MULTIROIR
Laboratory trays, coloured
Katalog # 216-1130
Leverandør:  MULTIROIR
Laboratory trays, coloured
Laboratory trays, coloured
rød, 260×196×42 mm, 210×150×40 mm
Price
108,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PS.
  • Five different colours
  • Two sizes
  • Good chemical and temperature resistance
Multipurpose trays for a wide range of applications.
Specifikationer
  • B×D×H udv.:
    260×196×42 mm
  • Farve:
    rød
  • Seq:
    10
  • B×D×H indv.:
    210×150×40 mm