Calibrated Disposable Plastic Inoculation Loops
Rigid (hard) plastic inoculation loop, 10 µl, mørkeblå, 20/pouch
: Copan
- Pakket:20/pouch
- Kapacitet:10 µl
- Farve:mørkeblå
- Beskrivelse:Rigid (hard) plastic inoculation loop
- Seq:06