Orders LinkContactUsLinkComponent
Calibrated Disposable Plastic Inoculation Loops
Rigid (hard) plastic inoculation loop, 10 µl, mørkeblå, 20/pouch
  710-0198
 :   Copan
Calibrated Disposable Plastic Inoculation Loops
  710-0198
 :  Copan
Calibrated Disposable Plastic Inoculation Loops
Rigid (hard) plastic inoculation loop, 10 µl, mørkeblå, 20/pouch
562,00 kr
1 * 1.000 Items
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
  • Pakket:
    20/pouch
  • Kapacitet:
    10 µl
  • Farve:
    mørkeblå
  • Beskrivelse:
    Rigid (hard) plastic inoculation loop
  • Seq:
    06