Calibrated Disposable Plastic Inoculation Loops
Rigid (hard) plastic inoculation loop, 10 µl, mørkeblå, 10/pouch
Supplier: Copan
Jump to:
Product Details & Documents
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
Documents
Certificates
Search by Lot NumberSpecifications
- Pakket:10/pouch
- Kapacitet:10 µl
- Farve:mørkeblå
- Beskrivelse:Rigid (hard) plastic inoculation loop
- Seq:11