Instrument boxes
sølvfarvet, 100×215×30 mm
Catalog # AGEN8034
Supplier: Agencinox
Jump to:
Product Details & Documents
Aluminium, anodised.
- Smooth surfaces for fast, easy cleaning
- Coloured
- Good chemical resistance, very good thermal resistance
Specifications
- Farve:sølvfarvet
- Seq:04
- B×D×H:100×215×30 mm
- Cat. No.:AGEN8034