Orders LinkContactUsLinkComponent
Instrument boxes
Instrument boxes
sølvfarvet, 90×180×30 mm
Katalog # AGEN8030
Leverandør:   Agencinox
Instrument boxes
Katalog # AGEN8030
Leverandør:  Agencinox
Instrument boxes
Instrument boxes
sølvfarvet, 90×180×30 mm
Price
267,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Aluminium, anodised.
  • Smooth surfaces for fast, easy cleaning
  • Coloured
  • Good chemical resistance, very good thermal resistance
Specifikationer
  • Farve:
    sølvfarvet
  • Seq:
    02
  • B×D×H:
    90×180×30 mm