Orders LinkContactUsLinkComponent
Accessories for centrifuges
Accessories for centrifuges
Adapter for 10×1,5/2,0 ml microtube, 10,8×37 mm
  521-1305
   Lav fremstillingspåvirkning
 :   Hettich
Accessories for centrifuges
  521-1305
 :  Hettich
   Lav fremstillingspåvirkning
Accessories for centrifuges
Accessories for centrifuges
Adapter for 10×1,5/2,0 ml microtube, 10,8×37 mm
509,00 kr
1 * 1 Items
  • Beskrivelse:
    Adapter for 10×1,5/2,0 ml microtube, 10,8×37 mm
  • Seq:
    02
  • Cat. No.:
    521-1305
  • Miljøcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning