Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Halvmasker, Valuair

Economy, M
Katalog # 111-3520
Leverandør:   HONEYWELL SAFETY
Halvmasker, Valuair
Katalog # 111-3520
Leverandør:  HONEYWELL SAFETY
Halvmasker, Valuair
Halvmasker, Valuair

Economy, M

Price
231,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Reusable half masks.
Economy: Silicone-free half masks, with a texture that combines comfort and exceptional impermeability for facial protection.
Premier: Half mask made from supple silicone for good flexibility, comfort and resistance; perspiration evacuation hole to prevent skin irritation and simplified fastening mechanism.

  • Dobbeltsidet filtersystem sikrer forbedret udsyn
  • Nem, sikker tilpasning med 'click-fit'-mekanisme

Filtertilslutning: Bajonet

Economy: 109 g; Premier: 140 g

Filtertilslutning: Bajonet

Let: 109 g (størrelse M)

 : EN 140

 : Valuair half masks are designed for use with the Sperian range of filter cartridges (111-3540, 111-3528, 111-3529, 111-3530, 111-0247).

Specifikationer
  • Model:
    Economy
  • Seq:
    01
  • Størrelse:
    M