Orders LinkContactUsLinkComponent
Dissecting trays
Dissecting trays
500×350×50 mm
Katalog # NICK698
Leverandør:   NICKEL ELECTRO
Dissecting trays
Katalog # NICK698
Leverandør:  NICKEL ELECTRO
Dissecting trays
Dissecting trays
500×350×50 mm
Price
1.090,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy handling
For use in laboratories and schools.
Specifikationer
  • B×D×H:
    500×350×50 mm