96-Multiwell insert system, Falcon®, Corning
Fem indsatsplader med fødebakker og låg
Katalog # 734-0368
Leverandør: Corning
96-Multiwell insert system, Falcon®, Corning
Katalog # 734-0368
Leverandør: Corning
Gå til:
Product Details & Documents
The Falcon® HTS 96-multiwell insert system is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies. This automation compatible platform is composed of a 1,0 µm pore size PET membrane-based 96-multiwell insert plate, a media feeder tray and a lid. The drop-in baffle for the feeder tray mitigates media sloshing and lowers the risk of contamination.
- Automation compatible design suitable for use with most robots and fluid handling instruments
- Excellent reproducibility - the one-piece feeder tray enhances consistency in well-to-well monolayer growth
- Ideal for drug transport studies
Specifikationer
- Beskrivelse:Fem indsatsplader med fødebakker og låg
- Cat. No.:734-0368
- Miljøcertificering:ISO 14001 - Bedford
- Environmentally Preferable:Lav fremstillingspåvirkning
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- 96-square well, angled-bottom plate, with lid96-square well, angled-bottom plate, with lid734-1378Log ind for tilgængelighed1 * 5 Items - 4.280,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Fødebakke, PS, ikke behandlet, designet til brug sammen med Falcon® 96-multibrønds indsatssystemerFødebakke, PS, ikke behandlet, designet til brug sammen med Falcon® 96-multibrønds indsatssystemer736-2024Log ind for tilgængelighed1 * 5 Items - 1.510,00 kr