Orders LinkContactUsLinkComponent
Retort clamps
120 mm, 180 mm
  BOCH5481
 :   BOCHEM
Retort clamps
  BOCH5481
 :  BOCHEM
Retort clamps
Retort clamps
120 mm, 180 mm
323,00 kr
1 * 1 Items
Steel, zinc plated.
  • Round jaws with cork lining to protect clamped items
  • Without bosshead for fixing with stand
  • With wing screws for rapid easy clamping
DIN 12894
 : DIN 12894
  • Spændvidde:
    120 mm
  • Længde:
    180 mm
  • Seq:
    03