Retort clamps
120 mm, 180 mm
BOCH5481
: BOCHEM
Steel, zinc plated.
- Round jaws with cork lining to protect clamped items
- Without bosshead for fixing with stand
- With wing screws for rapid easy clamping
DIN 12894
: DIN 12894
- Spændvidde:120 mm
- Længde:180 mm
- Seq:03