Orders LinkContactUsLinkComponent
Retort clamps
Retort clamps
80 mm, 180 mm
Katalog # BOCH5464
Leverandør:   BOCHEM
Retort clamps
Katalog # BOCH5464
Leverandør:  BOCHEM
Retort clamps
Retort clamps
80 mm, 180 mm
Price
230,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Steel, zinc plated.
  • Round jaws with cork lining to protect clamped items
  • Without bosshead for fixing with stand
  • With wing screws for rapid easy clamping
DIN 12894
 : DIN 12894
Specifikationer
  • Spændvidde:
    80 mm
  • Længde:
    180 mm
  • Seq:
    01