Buretteklemmer
Steel, zinc plated, fingers with silicone coating, 0 - 20 mm, To buretter
Katalog # BOCH5477
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Burette clamps made of zinc plated stainless steel.
- Fingers are covered with silicone coating to protect clamped items
- Single sided or double sided available
Specifikationer
- Til:To buretter
- Spændvidde:0 - 20 mm
- Materiale:Steel, zinc plated, fingers with silicone coating