Orders LinkContactUsLinkComponent
Buretteklemmer
Buretteklemmer
Steel, zinc plated, fingers with silicone coating, 0 - 20 mm, En burette
Katalog # BOCH5476
Leverandør:   BOCHEM
Buretteklemmer
Katalog # BOCH5476
Leverandør:  BOCHEM
Buretteklemmer
Buretteklemmer
Steel, zinc plated, fingers with silicone coating, 0 - 20 mm, En burette
Price
351,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Burette clamps made of zinc plated stainless steel.
  • Fingers are covered with silicone coating to protect clamped items
  • Single sided or double sided available
Specifikationer
  • Til:
    En burette
  • Spændvidde:
    0 - 20 mm
  • Materiale:
    Steel, zinc plated, fingers with silicone coating