Klemmer, mikro
Clamps, micro, finger with silicone coating, Rustfrit stål 18/10, 0 - 20, 150 mm
Katalog # BOCH5415
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Micro clamps made of stainless steel.
- Fingers are covered with either a layer of cork or with silicone coating to protect clamped items
- With round or angled jaws
Specifikationer
- Spændvidde:0 - 20
- Beskrivelse:Clamps, micro, finger with silicone coating
- Længde:150 mm
- Materiale:Rustfrit stål 18/10