Vejeskeer
Vejeskeer, konisk form, 290 mm, 75 mm
Katalog # BOCH3605
Leverandør: BOCHEM
Gå til:
Product Details & Documents
These conical weighing scoops are made of 18/10 stainless steel.
Specifikationer
- Bredde:75 mm
- Beskrivelse:Vejeskeer, konisk form
- Længde:290 mm
- Seq:02