Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mohr pinch cocks

Mohr pinch cocks

15 mm, 50 mm
Katalog # 229-0064
Leverandør:   BOCHEM
Mohr pinch cocks
Katalog # 229-0064
Leverandør:  BOCHEM
Mohr pinch cocks
Mohr pinch cocks

15 mm, 50 mm

Price
29,70 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Nickel-plated brass.

Specifikationer
  • Spændvidde:
    15 mm
  • Længde:
    50 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.