Sealing tapes
Sealing tape, PetriSEAL™, hvid, 13,0 mm
Katalog # DIVEPSEAL-108W
Leverandør: DIVERSIFIED BIOTECH
Gå til:
Product Details & Documents
Sealing tapes in different colours.
- Different sizes
- Temperature resistant
- Chemically resistant
Length: 2,7 m
Specifikationer
- Bredde:13,0 mm
- Farve:hvid
- Beskrivelse:Sealing tape, PetriSEAL™
- Seq:04