Sealing tapes
Sealing tape, ContaineriSEAL™, hvid, 19,0 mm
Katalog # DIVECSEAL-75W
Leverandør: DIVERSIFIED BIOTECH
Gå til:
Product Details & Documents
Sealing tapes in different colours.
- Different sizes
- Temperature resistant
- Chemically resistant
Length: 2,7 m
Specifikationer
- Bredde:19,0 mm
- Farve:hvid
- Beskrivelse:Sealing tape, ContaineriSEAL™
- Seq:07