SKRUELÅG C4015-1A T/VIAL 100STK 1 * 100 Items
Katalog # NSCAC4015-1A
Leverandør: THERMO NATIONAL SCIENTIFIC
SKRUELÅG C4015-1A T/VIAL 100STK 1 * 100 Items
Katalog # NSCAC4015-1A
Leverandør: THERMO NATIONAL SCIENTIFIC
Frequently Bought Together
Vand, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverandør: VWR Chemicals
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Leverandør: VWR Chemicals
VWR®, Short Thread Vials ND9
Leverandør: VWR Collection
VWR®, volumetriske kolber, klasse A, glas, klar, NS plastikprop
Leverandør: VWR Collection
Ethanol 70% ≥70% (v/v), TechniSolv®
Leverandør: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Leverandør: VWR Collection
Vand, LiChrosolv® for chromatography, for LC-MS, Supelco®
Leverandør: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
VWR®, Ansigtsservietter
Leverandør: VWR Collection
VWR®, Nitril handsker, NITRILE
Leverandør: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals