Orders LinkContactUsLinkComponent
Cream butyrometer, without stopper
Cream butyrometer, without stopper
Butyrometer for cream, 0 - 20 % : 0,2
Katalog # 614-0106
Leverandør:   GERBER FUNKE
Cream butyrometer, without stopper
Katalog # 614-0106
Leverandør:  GERBER FUNKE
Cream butyrometer, without stopper
Cream butyrometer, without stopper
Butyrometer for cream, 0 - 20 % : 0,2
Price
167,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Measuring method, for ice cream.
Specifikationer
  • Beskrivelse:
    Butyrometer for cream, 0 - 20 % : 0,2
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Stopper, rubber, conical, for all measuring method butyrometers
    Stopper, rubber, conical, for all measuring method butyrometers
    614-2028
    Log ind for tilgængelighed
    1 * 100 Items - 360,00 kr