Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Volumetric flasks, class B, Azlon®

Volumetric flasks, class B, Azlon®

100 ml, 14/23, 0,20 ± ml
Katalog # 214-0117
Leverandør:   SciLabware
Volumetric flasks, class B, Azlon®
Katalog # 214-0117
Leverandør:  SciLabware
Volumetric flasks, class B, Azlon®
Volumetric flasks, class B, Azlon®

100 ml, 14/23, 0,20 ± ml

Price
207,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

PP, translucent, with PE stopper.

  • Class B tolerances with a clear easy to read calibration line
  • Feature a mixing chamber at the top of each flask's neck
  • Exposure to temperatures above 60 °C may affect accuracy
  • Cleaning with mild alkaline detergents recommended up to 60 °C

Certifications: ISO 1042

Specifikationer
  • NS:
    14/23
  • Kapacitet:
    100 ml
  • Tolerance:
    0,20 ± ml