Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux, Quickfit®
Fractionating columns, Vigreux, Quickfit®
250 mm, 19/26 NS, 19/26 NS
Catalog # 201-8243
Fractionating columns, Vigreux, Quickfit®
Catalog # 201-8243
Supplier:  
Fractionating columns, Vigreux, Quickfit®
Fractionating columns, Vigreux, Quickfit®
250 mm, 19/26 NS, 19/26 NS
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
GLASKOLONNE VIGREUX M/KAPPE NS19 200MM 1 * 1 Items Catalog #201-0175
Product 201-0175 is similar to 201-8243 but may have a different manufacturer and/or packaging.
Product Details & Documents
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
No glass jacket. With combination of horizontal and inclined indentations. Horizontal indentations ensure intimate vapour and piquid contatc. Inclined indentations redistribute liquid from walls to centre of column.
Specifications
  • Konusstr.:
    19/26 NS
  • Kappestr.:
    19/26 NS
  • Brugbar længde:
    250 mm
  • Seq:
    01