Adapters, receiver, vacuum, Quickfit®
29/32 NS, 29/32 NS, 165 mm
Katalog # 201-0599
Leverandør: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, bent.
- Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.
Specifications
- Konusstr.:29/32 NS
- Kappestr.:29/32 NS
- Samlet længde:165 mm