Orders LinkContactUsLinkComponent
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
29/32 NS, 29/32 NS, 165 mm
Katalog # 201-0599
Leverandør:   SciLabware
Adapters, receiver, vacuum, Quickfit®
Katalog # 201-0599
Leverandør:  SciLabware
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
29/32 NS, 29/32 NS, 165 mm
Price
840,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, bent.
  • Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.
Specifications
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    29/32 NS
  • Samlet længde:
    165 mm