Knife spatulas, angled
230 mm, 130 mm
Katalog # HAMMHSN237-13
Leverandør: HAMMACHER KARL
Gå til:
Product Details & Documents
Flexible knife spatulas with wooden handle and blade made of stainless steel.
Specifikationer
- Længde:230 mm
- Seq:16
- Klinge længde:130 mm