Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Knife spatulas, angled

Knife spatulas, angled

205 mm, 105 mm
Katalog # HAMMHSN226-10
Leverandør:   HAMMACHER KARL
Knife spatulas, angled
Katalog # HAMMHSN226-10
Leverandør:  HAMMACHER KARL
Knife spatulas, angled
Knife spatulas, angled

205 mm, 105 mm

Price
493,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Flexible knife spatulas with wooden handle and angled blade made of stainless steel.

Specifikationer
  • Længde:
    205 mm
  • Seq:
    12
  • Klinge længde:
    105 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.