Orders LinkContactUsLinkComponent
Knife spatulas, angled
180 mm, 80 mm
  HAMMHSN225-08
Knife spatulas, angled
  HAMMHSN225-08
Knife spatulas, angled
Knife spatulas, angled
180 mm, 80 mm
492,00 kr
1 * 1 Items
Flexible knife spatulas with wooden handle and angled blade made of stainless steel.
  • Længde:
    180 mm
  • Seq:
    11
  • Klinge længde:
    80 mm