Orders LinkContactUsLinkComponent
Instrument dishes
Instrument dishes
Låg til 216-1401 skåle, 120x60x10 mm
Katalog # 216-1411
Instrument dishes
Katalog # 216-1411
Instrument dishes
Instrument dishes
Låg til 216-1401 skåle, 120x60x10 mm
Price
172,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Thick-walled glass.
  • Can be sterilised in hot air up to 180 °C
  • Robust
  • Stainless steel lids are available separately
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Specifikationer
  • Beskrivelse:
    Låg til 216-1401 skåle
  • Seq:
    08
  • B×D×H:
    120x60x10 mm