Orders LinkContactUsLinkComponent
Quality control test suspensions
Bacillus subtilis, 10 mL
Katalog # NAMSGP-01
Leverandør:   CROSSTEX
Quality control test suspensions
Katalog # NAMSGP-01
Leverandør:  CROSSTEX
CAS nummer:  
Quality control test suspensions
Bacillus subtilis, 10 mL
CAS nummer:  
Price
2.830,00 kr
Log ind for tilgængelighed
1 * 10 mL
Product Details & Documents
The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.
  • 10 ml pro vial
  • 100 dose
Documents
Specifikationer
Specifikationer
  • Pakn:
    10 mL
  • Beskrivelse:
    Bacillus subtilis