Orders LinkContactUsLinkComponent
Staining plates
Staining plates
10, Ø×D: 14×2 mm, 105×55×5 mm
Katalog # HECH40623010
Staining plates
Katalog # HECH40623010
Staining plates
Staining plates
10, Ø×D: 14×2 mm, 105×55×5 mm
Price
343,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Glass spot plates with 10 or 12 numbered cavities. For flocculation tests.
Specifikationer
  • Ant. brønde:
    10
  • Seq:
    01
  • B×D×H:
    105×55×5 mm
  • Brøndstørrelse:
    Ø×D: 14×2 mm