Calibrated Disposable Plastic Inoculation Loops
Flexible (soft) plastic inoculation loop, 10 µl, lyseblå, 20/pouch
Leverandør: Copan
Gå til:
Product Details & Documents
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Pakket:20/pouch
- Kapacitet:10 µl
- Farve:lyseblå
- Beskrivelse:Flexible (soft) plastic inoculation loop
- Seq:04