Flexible (soft) plastic inoculation loop, 1 µl, lysegrøn, 20/pouch
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
#: HIRS9221275
: HIRSCHMANN
#: 631-0124
: VWR Collection
#: 710-0449
: Copan
#: 710-0432
#: 612-5755
#: 612-2851
#: 1.10962.0003
: Merck
#: 291-0057
: HEATHROW