Calibrated Disposable Plastic Inoculation Loops
Flexible (soft) plastic inoculation loop, 1 µl, lysegrøn, 10/pouch
Supplier: Copan
Jump to:
Product Details & Documents
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
Documents
Certificates
Search by Lot NumberSpecifications
- Pakket:10/pouch
- Kapacitet:1 µl
- Farve:lysegrøn
- Beskrivelse:Flexible (soft) plastic inoculation loop
- Seq:07