Orders LinkContactUsLinkComponent
Calibrated Disposable Plastic Inoculation Loops
Calibrated Disposable Plastic Inoculation Loops
Flexible (soft) plastic inoculation loop, 1 µl, lysegrøn, 10/pouch
Catalog # 710-0191
Supplier:   Copan
Calibrated Disposable Plastic Inoculation Loops
Catalog # 710-0191
Supplier:  Copan
Calibrated Disposable Plastic Inoculation Loops
Flexible (soft) plastic inoculation loop, 1 µl, lysegrøn, 10/pouch
Price
1.310,00 kr
Log in for availability
1 * 2.000 Items
Product Details & Documents
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
Documents
Specifications
  • Pakket:
    10/pouch
  • Kapacitet:
    1 µl
  • Farve:
    lysegrøn
  • Beskrivelse:
    Flexible (soft) plastic inoculation loop
  • Seq:
    07