Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sølvnitrat ≥99.5%, GPR RECTAPUR®

Sølvnitrat ≥99.5%, GPR RECTAPUR®

Plastic bottle for solids, 100 g
Catalog # 21570.182
Oxidizing
Irritant
Corrosive
Environment
Supplier:   VWR Chemicals
Sølvnitrat ≥99.5%, GPR RECTAPUR®
Catalog # 21570.182
Supplier:  VWR Chemicals
CAS Number:  7761-88-8
Sølvnitrat ≥99.5%, GPR RECTAPUR®
Sølvnitrat ≥99.5%, GPR RECTAPUR®

Plastic bottle for solids, 100 g

CAS Number:  7761-88-8
Price
3.700,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:210 °C
  • Density:4,35 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Foreign heavy metals (as Pb):
    Max. 20 ppm
  • Not precipitated by HCl (as SO4):
    Max. 100 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 10 ppm
  • Pb (Lead):
    Max. 20 ppm

Specifications

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    100 g

Specifications