Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Ammoniumnitrat ≥98.5%, GPR RECTAPUR®

Plastic bucket, 5 kg
  21278.364
 
Ammoniumnitrat ≥98.5%, GPR RECTAPUR®
  21278.364
 :  6484-52-2
Ammoniumnitrat ≥98.5%, GPR RECTAPUR®
Ammoniumnitrat ≥98.5%, GPR RECTAPUR®

Plastic bucket, 5 kg

 :  6484-52-2
 
1.640,00 kr
   
1 * 5 kg
 
  •  :6484-52-2
  •  :NH₄NO₃
  •  :MFCD00011425
  •  :80.04 g/mol
  •  :13,00538
  •  :230 °C (1013 hPa)
  •  :169 °C
  •  :1,73 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :229-347-8
  •  :1942
  •  :5.1,III
 

 

  • Assay (calculated on anhydrous):
    Min. 98.5 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 10 ppm

 

  • Pakning:
    Plastic bucket
  • Pakn:
    5 kg

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.