Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Ammoniumnitrat ≥98.5%, GPR RECTAPUR®

Plastic bottle for solids, 1 kg
Katalog # 21278.295
Oxiderende
Leverandør:   VWR Chemicals
Ammoniumnitrat ≥98.5%, GPR RECTAPUR®
Katalog # 21278.295
Leverandør:  VWR Chemicals
CAS nummer:  6484-52-2
Ammoniumnitrat ≥98.5%, GPR RECTAPUR®
Ammoniumnitrat ≥98.5%, GPR RECTAPUR®

Plastic bottle for solids, 1 kg

CAS nummer:  6484-52-2
Price
388,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
  • CAS nummer:6484-52-2
  • Formel:NH₄NO₃
  • MDL nummer:MFCD00011425
  • Molecular Weight:80.04 g/mol
  • Merck Index:13,00538
  • Kogende Pt:230 °C (1013 hPa)
  • Smeltende Pt:169 °C
  • Tæthed:1,73 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:229-347-8
  • FN:1942
  • ADR:5.1,III
Specifikationer

Specifikationstestresultater

  • Assay (calculated on anhydrous):
    Min. 98.5 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 10 ppm

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    1 kg

Specifikationer